Gold, disc, self cleaning foil disc apertures 2 mm O.D., Thickness 0.53 mm

Supplier: Electron Microscopy Sciences

42015-AU 42020-AU 42030-AU 42050-AU 42100-AU 42005-AU 42070-AU 42040-AU 42010-AU
100496-628EA 455.26 USD
100496-628 100496-632 100496-636 100496-644 100496-648 100496-622 100496-646 100496-640 100496-626
Gold, disc, self cleaning foil disc apertures 2 mm O.D., Thickness 0.53 mm
Gold

Advanced techniques of thin metal film application permits the apertures to retain a clean hole edge during prolonged beam exposure


Gold foil aperture tolerances vary as follow:
• Nominal hole size from 1 to 60 micron: +/-1micron
• Nominal hole size 70 micron and up: +/-10%


Ultra-thin, half-micron design discourages contaminant accumulation and stabilizes astigmatic conditions to provide longer life. Conventional mechanical cleaning involving costly down-time is no longer necessary. Because the foil is ultra thin, care must be taken when handling the gold foil apertures.

Formula: Au
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR