Evaporated Platinum/Iridium on Holey Carbon Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80020
100496-366EA 111.87 USD
100496-366
Evaporated Platinum/Iridium on Holey Carbon Film, Electron Microscopy Sciences
Graticules, Reticles and Grids
Evaporated Pt/Ir on Holey carbon film.
The dots of evaporated Pt/Ir provide dense particles for resolution checks through the particle seperation test.
Holey carbon film support provides holes for ease of focus and astigmatism correction.