Zerostat Anti-Static Gun, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60610
100496-120EA 183.08 USD
100496-120
Zerostat Anti-Static Gun, Electron Microscopy Sciences
Surface Treatments Antistatic Treatments
The Zerostat Anti-Static Gun operates on a piezoelectric effect to emit positive and negative charges to neutralize static on any surface. This anti-static gun emits no radioactivity and does not require electricity or batteries.
Measures: 4×6"