Neon Bulb Array, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
97036-01
100495-474EA 219.36 USD
100495-474
Neon Bulb Array, Electron Microscopy Sciences
Lights Neon Lamps
This Neon Bulb Array is made from silicone
Each bulb in the mat is 2.5 cm apart. The mat is divided into quadrants. A mark on the back left corner of the array lid is used to key the back left corner of the oven. There are letters and numbers along each edge for easy placement on the Alpha-numeric tray. The mat size is 8 x 8".