Rectangular and Square Cover Slips (Bellco Glass), Electronic Microscopy Sciences
Supplier: Electron Microscopy Sciences
72190-35 72190-50 72192-75 72192-54 72191-50 72192-50 72190-09
100493-652EA 124.17 USD
100493-652 100493-654 100493-664 100502-206 100493-656 100493-660 100493-650 100493-662 100493-648
Rectangular and Square Cover Slips (Bellco Glass), Electronic Microscopy Sciences
Cover Glasses
These cover slips provides good quality, reliability, and better performance for all laboratory microscopy needs
  • #1 thickness (0.13 to 0.16 mm)
  • Packed one ounce (28 g) per case
  • Clear white borosilicate glass.
For use with Bellco Leighton Tissue Culture Tubes.