Scalpel Handles Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72040-08 72040-05 72040-06
100502-834EA 121.41 USD
100502-834 100493-600 100493-602
Scalpel Handles Disposable, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpel Handles
These strong, moulded plastic scalpel handles provide a secure palm-fitting grip for post-mortem work, trimming light plaster casts, and similar forceful cutting.
Handle size #6 and #8 Bard-Parker* brand.