Mini Vacuum Desiccator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71246-01
100493-228EA 168.68 USD
100493-228
Mini Vacuum Desiccator, Electron Microscopy Sciences
Desiccators Desiccators, Plastic
The top part of the desiccator is clear, molded from polycarbonate and see-through from any angle. A polyethylene stopcock with a "PTFE" plug allows for gentle releasing of the vacuum. A peripheral flange around the polypropylene bottom section fits into and snap locks therefore sealing the two halves against a "O" ring. Vacuum retention rating is 740 mm Hg (29"). Inside dimensions are 102mm (4") diameter x 59mm (2 7/16")high. The base is 175mm (6 7/8") diameter.
  • Ideal for Small Samples
  • Degassing Embedding mixtures