Replacement Stopcock, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71242-05
100493-220EA 110.5 USD
100493-220
Replacement Stopcock, Electron Microscopy Sciences
Stopcocks
A polypropylene body which has a TFE plug and washer as well as a bright red handle and nut.
Its taper fits on the desiccator port and the plug is 3-way allowing for vacuum draw, shut-off, and release. May be used with EMS vacuum desiccators.