Dry Keeper Plus™ Desiccator Cabinet; Non-Vacuum, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71218-00
100493-146EA 3867.19 USD
100493-146
Dry Keeper Plus™ Desiccator Cabinet; Non-Vacuum, Electron Microscopy Sciences
Desiccators Desiccator Cabinets
Efficient dehumidification is monitored by a hygrometer dial in the door
  • User-friendly electronic desiccant control
  • One-touch door latch with silicone rubber door gasket
  • Molded cabinet made from ABS plastic
Overall measurement: 310 x 353 x 430 mm (12.2 x 13.9 x 16.9")
Hinged door measures: 300 x 355 mm (11.8 x 14"). Weight: 5.9 kg (13lbs.)
The dehumidifying auto-drying unit has an air circulating system which helps drop relative humidity sharply through its cycling of absorption and regeneration of the enclosed permanent desiccant. It can bring the relative humidity down from 60% to 10% in 6 hours of continuous operation in an unloaded cabinet. The unit comes with three clear, removable shelves with vent holes which provide evenly spaced storage area. It operates on 110V, 60 Hz., 10 watts, and comes with a 2-prong 6 ft (2 meter) cord.