Glass Knife Boat: Quick Boat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71009-60 71009-10 71009-64 71009-80
100493-042EA 293.08 USD
100493-042 100493-040 100493-044 100493-046
Glass Knife Boat: Quick Boat, Electron Microscopy Sciences
Microtomes and Microtome Assemblies Microtome Blades and Handles
These boats are made from aluminum and are anodized black. They come complete with a set screw on one side.
The area where the glass knife will be attached, is cut to fit to the width of the knife and is cushioned by silicone rubber to ensure a tight seal and no leakage.