Slide Staining Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70321-20
100492-958EA 100.21 USD
100492-958
Slide Staining Dish, Electron Microscopy Sciences
Jars Staining Jars
The dish is made from polymethylpentene "glass clear" for use with our Polypropylene staining rack.
It comes with two lids; one lid should be used when the rack is not inserted.
Dimensions: 81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8").