Slide Staining Rack; Polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70321-10
100492-956EA 50.01 USD
100492-956
Slide Staining Rack; Polypropylene, Electron Microscopy Sciences
Staining Boxes
This slide staining rack is made from polypropylene and is autoclavable.
  • Accommodates up to 20 slides 76×26 mm (3×1")
  • Comes with a removable snap-on handle
  • Measures: 70×86×21 mm (2¾×3⅜×¹³⁄₁₆") (without handle)