Sapphire Blade/Knife, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
93060
100492-502EA 1197.78 USD
100492-502
Sapphire Blade/Knife, Electron Microscopy Sciences
Knives
A unique high-precision knife for use with all Vibratomes and Tissue Sectioners, as well as Oscillating Tissue Slicers (EMS 4000 and EMS 5000).
This Sapphire Knife offers smooth, consistent and thinner sections than are usually obtainable with stainless steel blades. The blade allows for sectioning as thin as 10 microns. Offering many years of cutting surface, the durability of our sapphire knife far outlasts other comparable blades on the market.
Material: Synthetic Single Crystal Sapphire
Size: 1.5 x 0.4 x .03”