Gold, sputter coater target, Diameter 60 mm, Thickness 0.1 mm

Supplier: Electron Microscopy Sciences

91010
100492-480EA 1271.9 USD
100492-480
Gold, sputter coater target, Diameter 60 mm, Thickness 0.1 mm
Gold

These sputter coater targets are intended for EMS equipment and are stock items. Targets for other coaters are custom made and delivery is about one week. We are able to supply targets (foil with base) for the following sputtering units: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences and others.


  • Available in Gold, Gold/Palladium, Platnium, Chromium, and Silver
  • Up to 30% thicker than the original


Ordering information: When ordering, please state make and model of the sputtering unit to ensure proper fit.

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR