Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74839
100492-264EA 142.8 USD
100492-264
Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Racks Draining Racks
For air drying up to 20 RC prints in 15 min. Its compact design permits maximum ambient air circulation and prevents trapping of water between the print and the rack. The rack is also suitable for use with fiber-based paper. Print capacity: 5- 11 x 14"; 10-8 x 10"; 20-5 x 7".