Thermostatic Dryer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74836
100492-262EA 293.25 USD
100492-262
Thermostatic Dryer, Electron Microscopy Sciences
Heaters Hot Air Dryers
Used for RC and fiber base papers.
  • Fast start
  • Quick drying
  • No over heating
  • Drying surface: 30.5×43.1 cm (12×17")
It accommodates one 11×14", two 8×10" or eight 4×5" prints at once on each drying surface.
Has maximum uniformity of surface heat. Made from a highly polished chrome Ferrotype metal.