Tantalum, foil, Thickness 0.003" (0.075 mm)

Supplier: Electron Microscopy Sciences

73822
100492-114EA 382.56 USD
100492-114
Tantalum, foil, Thickness 0.003" (0.075 mm)
Tantalum

Tantalum foil, 0.003" thick, (0.075mm), 6 x 6" (15.2 x 15.2 cm), for aperture cleaning, vacuum evaporation.

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
MDL Number: MFCD00011252
CAS Number: 7440-25-7
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR