Microbeaker Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72645 72645-10
100491-988EA 83.42 USD
100491-988 100493-824
Microbeaker Holder, Electron Microscopy Sciences
Racks Bottle Racks
Brushed aluminum rack holds six 12 mm O.D. microbeakers in unique, stepped "see-through" holes for illuminated and microscopic observation of samples.