Vial Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72640
100491-986EA 82.92 USD
100491-986
Vial Holder, Electron Microscopy Sciences
Racks Chromatography Vial Racks
Machined from solid aluminum, holds 5 shell vials. It is ideal for handling cold or hot samples for prolonged periods of time out of the refrigerator or oven. Comes complete with a handle. Measurements: 4 3/4 x 1 1/4 x 3/4".