PTFE Surface Protectors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72612
100491-974EA 546.85 USD
100491-974
PTFE Surface Protectors, Electron Microscopy Sciences
Surface Protectors Bench Protectors
A thickness of 0.001" "PTFE" film bonded to a 0.008" self-adhesive vinyl gives your work surface good protection.
Easy to use: cut to desired size, remove the backing, and press into place.