Low Intensity UV Lamp, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72422 72425 72423 72424
100491-958EA 519.74 USD
100491-958 100491-964 100491-960 100491-962
Low Intensity UV Lamp, Electron Microscopy Sciences
Lights UV Lights, Lamps and Bulbs
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements.
Constructed from Cyolac® plastic, with a longwave emission of 365 nm and an intensity at 6" (15.2 cm) of 750 µm/cm². The bulb has a rated life of 6000 hours. It can be used as a hand-held light or it may br mounted on a optional stand. Powered by 110 V/60 Hz or 220 V/50 Hz.