Illuminated Slide Viewing Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71660
100491-814EA 77.72 USD
100491-814
Illuminated Slide Viewing Box, Electron Microscopy Sciences
Lights Light Boxes
Sorting and viewing 2×2" slides easily and quickly is made possible with this slide sorter unit. All metal frame, its baked white enamel finish reflects light for perfect viewing of slides.
  • Translucent styrene viewing panel
  • Folds flat for storage
  • Uses standard 60-watt household bulbs
Model S-40 Holds 40 slides sized 12×16". Viewing surface 30.4×40.6 cm.
Packaging: Bulb not included.