Micro-Test Staining Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71564
100491-792EA 98.51 USD
100491-792
Micro-Test Staining Dish, Electron Microscopy Sciences
Jars Staining Jars
This staining dish is made from clear glass and has 10 cells in 2 rows of 5 each. Each cell is 2 mm deep and holds 0.15 ml of solution.
Very useful in specimen preparation, EM staining, and Boerner-Jones-Lukens microfluocculation test.
Measures 108 X 57 mm (4¼" x 2¼").