Low Temperature Dessicator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71222 71220-A 71220 71222-A 71221 71221-A
100491-762EA 1176.99 USD
100491-762 100493-162 100502-804 100493-170 100491-760 100493-164
Low Temperature Dessicator, Electron Microscopy Sciences
Desiccators Desiccator Cabinets
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm (1/4") acrylic with full-length hinges and stainless steel twist locking latches.
  • Easy viewing clear units or dark bonze for light sensitive materials
  • Deep, slide-in trays with perforated bottom
  • Available in three sizes