Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70624
100491-640EA 55.21 USD
100491-640
Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Pens Lab Markers
The Coat-Quick "G" pen improves the adherence of tissue sections onto the grids.
  • Used in pretreating grids prior to mounting supporting films such as formvar and carbon
  • Minimizes dislodging, widening, or breaking of the support film
With a touch of the pen to the grid, a thin layer of coating is applied to the grid. Drying takes place in approximately 1 to 2 minutes at room temperature. After it has dried the grid is ready for tissue mounting. The pen is also used in pretreating grids prior to mounting supporting films such as formvar and carbon; it minimizes dislodging, widening, or breaking of the support film.