EMS Immunofluorescence Chamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70330
100491-614EA 123.32 USD
100491-614 100491-616
EMS Immunofluorescence Chamber, Electron Microscopy Sciences
Culture Dishes Bioassay Dishes
A two well incubation chamber for use in immunocytochemistry. Also available in a one well version. This chamber accepts regular microscope slides or coverslips and is assembled without any bonding substrates. Antibody incubation and washing are done in the chamber wells. It is then taken apart before microscopic examination. It is a proven solution to contamination spills, non-specific fluorescent signals, and sample dehydration in indirect immunofluorescence assays.
  • Very reliable and will accept a variety of samples and antibody washing conditions
  • Drastically reduces non-specific backgrounds
  • Elimination of contamination