Slide Duplicating Molds, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70170
100491-608EA 39.19 USD
100491-608
Slide Duplicating Molds, Electron Microscopy Sciences
Moulds Moulds for Histology
It uses a 1×3" glass slide as a base, and duplicates the slide using the resin of your choice.
Tissue sections are embedded against the flat glass surface. This duplicated resin slide can be studied under the light microscope, then cut, remounted, resectioned, and studied under the EM. A three cavity mold, each cavity measuring: 1W×3³/₃L×2"D. Reference instructions enclosed.