Tissue Matrices, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69012 69016 69015 69010 69014 69011 69013
100491-530EA 1253.03 USD
100491-530 100491-538 100491-536 100491-526 100491-534 100491-528 100491-532
Tissue Matrices, Electron Microscopy Sciences
Specimen Mounting Supplies
The Tissue Matrix is designed to aid the basic research scientist in the free-hand dissection of a wide variety of different tissues and repeatedly slice fresh tissue at intervals as small as 1 mm.
  • Channels at 1mm intervals
  • Channel width is 0.3mm
  • Removable end caps
  • Available in 3 basic configurations
  • Overall L x W x H Measurements: 90 x 36 x 37 mm
The Tissue Matrix is designed to aid the basic research scientist in the free-hand dissection of a wide variety of different tissues and repeatedly slice fresh tissue at intervals as small as 1 mm. The tissue matrices are constructed from high grade aluminum to allow for rapid temperature equilibration, ease of cleaning or sterilization and withstand the rigors of daily use.