Steinheil Magnifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68147 68146 68145 68148
100491-462EA 99.49 USD
100491-462 100491-460 100491-458 100491-464
Steinheil Magnifier, Electron Microscopy Sciences
Magnifiers
The magnifier helps the user eliminate bulky and heavier conventional magnifiers.
A biconvex lens. Pocket size. The sliding frame is designed for the storage of the lens after use. It is good for the preliminary inspection of microscopic specimens. It is lightweight and has excellent chromatic correction with a reasonably wide field of view. Size: 39 x 24 x 22 mm. Average weight: 20 g.