Parasite Magnifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68110
100491-434EA 88.58 USD
100491-434
Parasite Magnifier, Electron Microscopy Sciences
Magnifiers
An improved version of the conventional magnifying glass.
A 20 mm diameter parasite-like lens is cemented on a large 75 mm diameter lens. Magnification of the large one is 2.5X, while the small lens area has the magnification of 5X. The magnified image is very sharp and clear. Weight: 111g