Aluminium oxide, powder 15.0 µm

Supplier: Electron Microscopy Sciences

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

50362-15
100491-282EA 34.01 USD
100491-282
Aluminium oxide, powder 15.0 µm
Aluminium oxide

These are abrasive powders that are prepared from aluminum oxide, offering excellent coarse and intermediate polishing, with fast results.

Formula: Al₂O₃
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR