White Porcelain Plate, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71562-01 71562-06
100501-442EA 62.16 USD
100501-442 100491-004
White Porcelain Plate, Electron Microscopy Sciences
Desiccators Desiccators, Plates
Twelve cavities on a white porcelain plate. Used for staining and color reactions. They measure L x W: 118 x 90 mm (4 5/8 x 3 1/2"). Cavity depth: 6.4 mm (1/4").