Dropping Bottles, Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64220-05 64220-10 64221-05 64221-10
100490-426PK 156.3 USD
100490-426 100490-428 100490-430 100490-432
Dropping Bottles, Glass, Electron Microscopy Sciences
Bottles Dropping and Dispensing Bottles
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper.
A partial turn of the ground glass stopper will seal the bottle.