Sample Vials In Mini-File®; Clear, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60996-02
100490-068PK 124.27 USD
100490-068
Sample Vials In Mini-File®; Clear, Electron Microscopy Sciences
Vials
These vials are made from Wheaton-33R a low extractable borosilicate glass.
  • Vials and caps can be autoclaved
Delivery information: The vials are supplied with a white (styrene-butadiene) rubber lined cap.
Packaging: Comes ten vials packed in a clean plastic thermoformed Mini-File and offers convenience in transporting, displaying and storing.