Cellulose Acetate Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50420-30
100490-026PK 71.87 USD
100490-026
Cellulose Acetate Films, Electron Microscopy Sciences
Film Processing Supplies
This acetate film soften in acetone and can be used for duplicating purposes
Acetate film which is wet with acetone can then be applied to a surface which has been wet with acetone. It is easily removed away from the object when dry. Acetate films come in 35 µm thickness.