Spec-Cloth, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50320-05 50320-15
100490-006PK 70.65 USD
100490-006 100490-008
Spec-Cloth, Electron Microscopy Sciences
Wipes Disposable Wipes and Towels Lens Papers
A long napped rayon flock with a plastic barrier.
This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Available with Pressure Sensitive Adhesive (PSA) with and without hole.