Silk, Electron Microscopy
Supplier: Electron Microscopy Sciences
50312-05
100490-000PK 103.21 USD
100490-000
Silk, Electron Microscopy
Wipes Reusable Cleaning Cloths Cloths for Delicate Tasks
100% pure woven silk for intermediate, final, or skid polishing on friable materials. Available with Plain Backing (PB) without hole.
  • Size: 10" x N/H, P/B