Specialty Satin Cloth Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77030-06 77030-12 77030-09 77030-08 77030-11 77030-05 77030-10 77030-01 77030-02 77030-03 77030-04
100502-750PK 151.85 USD
100502-750 100489-642 100489-636 100489-634 100489-640 100489-632 100489-638 100489-624 100489-626 100489-628 100489-630
Specialty Satin Cloth Labels, Electron Microscopy Sciences
Labels
Labels withstand dry ice, liquid gases, deep freezes, acids, oils, greases, salt water, fresh water, and soap.
  • Sticks to almost any surface or shape
  • Mark with pen, pencil, or felt marker
  • Supplied pre-cut on protective backing paper
Designed to adapt to a wide range of laboratory procedures.Their textured surface may be marked with pen, pencil, or felt marker. Pressure-sensitive adhesive sticks to almost any surface or shape. Each label is perforated making it easy to remove the label from the roll.These labels are available in a variety of sizes. Labels ⁵/₈" in width come in sheet form only; all other sizes come on rolls of 1000 labels.