Inoculating Loop/Needle, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72952-10
100489-540PK 36.7 USD
100489-540
Inoculating Loop/Needle, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Nichrome wire. An 8” aluminum handle with a 3” nichrome wire, 25 gauge. Loop O.D. is 4 mm; loop I.D. is 2.5 mm. Available with a loop or a needle.