Rinzl Plastic Cover Slips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72261-19 72261-40 72261-50 72261-100 72261-60 72261-18 72261-22 72261-23
100489-432PK 89.95 USD
100489-432 100489-438 100489-440 100501-312 100489-442 100489-430 100489-434 100489-436
Rinzl Plastic Cover Slips, Electron Microscopy Sciences
Cover Glasses
Made from clear vinyl plastic.
  • Unbreakable, will not scratch or cut fingers
  • Used in Biological and Medical labs, etc.
  • Thickness 0.01" or 0.28 mm
  • Refractive index of glass
  • Economical and half the price of glass
  • Not autofluorescent