Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71471-Y 71471-G 71471-R 71470-G 71470-W 71470-Y 71470-B 71470-R 71471-B 71471-W
100489-302PK 159.35 USD
100489-302 100489-296 100489-298 100501-432 100501-436 100501-438 100496-328 100501-430 100501-434 102097-534 102097-536
Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
Microscope Slide Boxes
Made from durable polystyrene (with heavy walls).
This material is unaffected by humidity and is thoroughly insect proof. Each box is secured with a rust-resistant nickel-plated clasp and hinge pin. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 100 3x1" or 25 x 75 mm microscope slides. available in 5 color coded boxes making the boxes easy to identify. Each box measures 222 x 172 x 35 mm (8.75 x 6.75 x 3.375").