Desiccators Canister, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71205-01 71205-05
100489-242PK 35.77 USD
100489-242 100489-244
Desiccators Canister, Electron Microscopy Sciences
Desiccators Desiccants
This small container offers more desiccant in less space. It is ship-proof and possesses greater drying capabilities. Designed for use in bottles and small containers. Perfect for protecting against potency loss, odor formation, color changes, caking, mildew and condensation.
  • Provides superior absorption
  • Remains dry at maximum saturation
  • Silica gel can absorb 15% of its weight in water vapor in two hours
  • Silica Gel-Desiccant is a fast and extra moisture absorbing agent
Capsule shell is made from polyethylene with very fine mesh top and bottom. The canister contains 1 gram silica gel.