Long Molybdenum Boat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
73818
100488-506PK 58.83 USD
100488-506
Long Molybdenum Boat, Electron Microscopy Sciences
Boats Weighing Boats
Long Molybdenum Boat of 0.05 mm (0.002") thickness and 75 mm long.
  • Trough measures 50 mm (L) x 5 mm (W) x 1 mm (D).