Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71550
100488-442PK 30.26 USD
100488-442
Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Slide Mailers
The microscope slide container holds 5 slides without contact with one another. Ideal for air conveyor transport system. Measures: 3 L x 5/8 W x 11/4" H (51 x 16 x 32 mm).