Petri Dish, Analyslide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70650
100488-402PK 268.81 USD
100488-402
Petri Dish, Analyslide, Electron Microscopy Sciences
Culture Dishes Petri Dishes
The analyslide is like a petri dish on a slide. Use it as an examination and storage container for 47 mm filters.
  • Rectangular base fits any microscope stage
  • Inner cover ring offers airtight security for the samples and it does not interfere with viewing
  • Ideal for drying coated grids and absorbing water from grids that are supporting sections