Coverslip Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64005-01 64005-05
100488-186RL 455.32 USD
100488-186 100490-398
Coverslip Tape, Electron Microscopy Sciences
Cover Glasses
Coverslip tape has the same clarity as glass and fits most cover-slipper instruments.
The tape is loaded on a spool with 140 mm diameter core. This roll of coverslip tape eliminates the need for manual cover-slipping and extended user contact with xylene.