Tween® 20 (Polysorbate), Aurion

Supplier: Electron Microscopy Sciences

Synonyms: Polyoxyethylene sorbitan monolaurate

25564
100487-942EA 43.13 USD
100487-942
Tween® 20 (Polysorbate), Aurion
Tween® 20 (Polysorbate)

TWEEN-20® is a non-ionic surfactant with hydrophobic alkyl side chains.


In an aqueous environment the components form micelles above the critical micel concentration (at approximately 0.07% in PBS). This is based on hydrophobic interaction. When other hydrophobic components (proteins, gold conjugates etc.) are present these components are likely to get embedded in micellar structures. This has several effects: hydrophobic (background) interactions arelargely suppressed, the added different molecules are slightly increased in size and previously existing hydrophobic complexes are broken up due to surface forces.

Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR