HyperSep™ SAX Strong Anion Exchanger SPE Plates
Supplier: Thermo Scientific
60300-567
10047-760EA 653.2 USD
10047-760
HyperSep™ SAX Strong Anion Exchanger SPE Plates
Microplates Solid Phase Extraction Plates (SPE)
Strong anion exchange sorbent for extraction of weak acids.
  • Particle size: 40 to 63 µm
  • Pore size: 61 Å
  • Surface area: 481 m²/g