HyperSep Dispersive SPE Multipacks, Thermo Scientific
10047-114EA 327.57 USD
10047-114 10047-116 10047-118 10047-120 10047-122 10047-124 10047-126 10047-128
HyperSep Dispersive SPE Multipacks, Thermo Scientific
Chromatography Sorbents SPE Sorbents
Convenient and effective approach for determining pesticide residues in fruit, vegetables and other foods.
QuEChERS reagents are provided in individual metalized pouches
Ordering information: Each pack of 50 pouches is supplied with 50 empty centrifuge tubes with plug seal caps.