HyperSep Universal Vacuum Manifold, Thermo Scientific
Supplier: Thermo Scientific
60104-230
10047-012EA 6104.51 USD
10047-012
HyperSep Universal Vacuum Manifold, Thermo Scientific
Manifolds
Allows processing of SPE and SLE cartridges (3,4 flangeless and 6 mL) or 96-well plates
Allows flexibility in sample processing.
Delivery information: Includes manifold, base/gauge, flask and stopper, tubing and spigots.